Nalgene™ Heavy-Duty Cylindrical LLDPE Tanks with Spigots
Nalgene™ Heavy-Duty Cylindrical LLDPE Tanks with Spigots
Actual product may vary
Nalgene™ Heavy-Duty Cylindrical LLDPE Tanks with Spigots
Nalgene Heavy-Duty Cylindrical LLDPE Tanks include a needle-type spigot installed for draw off. Spigot accepts 5/8 inch I.D. tubing. Tanks are offered in sizes from 19 to 208L.
Have Questions?
Catalog NumberCapacity (Metric)
FKS1060383256.8 L
Catalog number FKS10603832
Price (MYR)
3,729.00
EA
Add to cart
Capacity (Metric):
56.8 L
Price (MYR)
3,729.00
EA
Add to cart
Thermo Scientific™ Nalgene Heavy-Duty Cylindrical LLDPE Tanks include a needle-type spigot installed for draw off. This spigot accepts 5/8 inch I.D. tubing and are available in sizes ranging from 19L to 208L.
Features:
  • Needle Type Tank Spigot (Cat. No. 6421) installed to allow draw off
  • 5/8 (15.9mm) I.D.
  • Not autoclavable
Included:
Cover
Note:
Tanks used at elevated temperatures or with high specific-gravity liquids should be externally supported.
Caution:
Plastic tanks are generally subject to more severe conditions that plastic labware; exposure is constant, stresses are greater and different classes and concentrations of chemicals are involved. Please pay special attention to chemical compatibility.
Warranty:
90 days
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Caution:
Plastic tanks are generally subject to more severe conditions that plastic labware; exposure is constant, stresses are greater and different classes and concentrations of chemicals are involved. Please pay special attention to chemical compatibility.
Specifications
Diameter (English) Outer13 in.
Diameter (Metric) Outer33 cm
TypeTank with Spigot
MaterialLLDPE
Capacity (English)15 gal.
Capacity (Metric)56.8 L
Height (English)27 in.
Height (Metric)68.6 cm
No. per Case1
No. per Pack1
Unit SizeEA