Orion™ DuraProbe™ Epoxy Bodied Graphite 4-Cell Conductivity Electrodes
Orion™ DuraProbe™ Epoxy Bodied Graphite 4-Cell Conductivity Electrodes
Actual product may vary
Orion™ DuraProbe™ Epoxy Bodied Graphite 4-Cell Conductivity Electrodes
Orion™ DuraProbe™ Epoxy Bodied Graphite 4-Cell Conductivity Electrodes
Orion™ DuraProbe™ Epoxy Bodied Graphite 4-Cell Conductivity Electrodes
Orion™ DuraProbe™ Epoxy Bodied Graphite 4-Cell Conductivity Electrodes
Orion™ DuraProbe™ Epoxy Bodied Graphite 4-Cell Conductivity Electrodes
Measure the most challenging samples accurately and under a wide range of conditions with these conductivity electrodes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCell ConstantConnector TypesLength (English) Cable
FKS105920130.475 cm-18-pin Mini-DIN10 ft.
FKS102557720.475 cm-18-pin Mini-DIN5 ft.
Catalog number FKS10592013
Price (MYR)
8,788.00
EA
Add to cart
Cell Constant:
0.475 cm-1
Connector Types:
8-pin Mini-DIN
Length (English) Cable:
10 ft.
Price (MYR)
8,788.00
EA
Add to cart
The Thermo Scientific™ Orion™ Conductivity electrode 013005MD complies with SM 2510B; D1125-95, -99 (A) for Drinking water measurements and EPA 120.1 (rev 1982), SM 2510 B, ASTM D1125-95 (99) (A) for Wastewater measurements.
Features
Virtually eliminate fringe field interference errors with the advanced Thermo Scientific™ Orion™ DuraProbe™ 4-Cell Conductivity Probes.
These probes are durable and extremely accurate over a wide range of conditions. The 4-Cell design compensates for electrode fouling, cable length, connector resistance, polarization errors, and fringe field interference errors. The epoxy/graphite cell material is extremely durable and chemically resistant with these conductivity probes being ideal for high & standard conductivity samples or difficult samples such as wastewater, runoff water and mud.
DuraProbe 4-cell conductivity probes are compatible with Thermo Scientific™ Orion Star™ , Lab Star™, Pro Star™, Versa Star Pro™ bench and portable conductivity meters. Paired together the instruments can be used to generate accurate, repeatable readings for conductivity, resistivity, TDS and salinity.
Solutions required
Standards
011008         100 µS/cm conductivity/TDS standard, 5 x 60 mL bottles
01100910     147 µS/cm conductivity standard, 10 pouches
011007         1413 µS/cm conductivity/TDS standard, 5 x 60 mL bottles
01100710     1413 µS/cm conductivity/TDS standard, 10 pouches
011006         12.9 mS/cm conductivity/TDS standard, 5 x 60 mL bottles
01100610     12.9 mS/cm conductivity/TDS standard, 10 pouches
011005         111 mS/cm conductivity standard, 5 x 60 mL bottles
01100510     111 mS/cm conductivity standard, 10 pouches
Items included with
011006         12.9 mS/cm conductivity/TDS standard, 60 mL 1 bottle
011007         1413 µS/cm conductivity/TDS standard, 60 mL 1 bottle
Warranty period
24 months from date of purchase
Specifications
For Use With (Equipment)Star A112, Star A122, Star A212, Star A215, Star A222, Star A322, Star A325, Star A329, Versa Star and Versa Star Pro Conductivity Meters
Sensor Types4 Electrode Graphite Cell
Cell Constant0.475 cm-1
Type4-Electrode Conductivity Cell, Waterproof 8-Din Connector
Connector Types8-pin Mini-DIN
DescriptionOrion DuraProbe Epoxy Bodied Graphite 4-Cell Conductivity Electrode
Diameter (English) Body0.59 in.
Diameter (Metric) Body15 mm
Length (English) Cable10 ft.
Length (Metric) Cable3 m
MaterialEpoxy
Measurement Ranges1 μS/cm to 200 mS/cm
Min. Immersion35 mm
Storage MethodStore in distilled or deionized water between measurements; for overnight or long-term storage, thoroughly rinse and store dry
ProbeConductivity probe with built-in ATC temperature sensor
Unit SizeEA