Class B Polypropylene Tall Form Measuring Cylinders
Class B Polypropylene Tall Form Measuring Cylinders
Actual product may vary
Class B Polypropylene Tall Form Measuring Cylinders
Manufactured from high-clarity polypropylene. Class B Polypropylene Tall Form Measuring Cylinders with blue printed scale provide easy volume assessment. Exposure to temperatures up to 80°C will not cause permanently exceeded error limits. Cleaning up to 60°C is recommended to preserve marks and inscriptions.
Have Questions?
Catalog NumberCapacity (Metric)Graduations
FKS105573912000 mL20 mL
Catalog number FKS10557391
Price (MYR)
494.00
EA
Add to cart
Capacity (Metric):
2000 mL
Graduations:
20 mL
Price (MYR)
494.00
EA
Add to cart
  • Manufactured from high-clarity polypropylene.
  • Tall form with blue printed scale provides easy volume assessment.
  • Class B.
  • Complies with DIN 12681 and ISO 6706 standards.
  • Calibrated to contain (TC, In.)
  • Exposure to temperatures up to 80°C will not cause permanently exceeded error limits.
  • Cleaning up to 60°C is recommended to preserve marks and inscriptions.
Specifications
Capacity (Metric)2000 mL
MaterialPolypropylene
Tolerance±20 mL
Certifications/ComplianceDIN 12681, ISO 6706
No. per Pack1 Pack
ClassB
CalibrationTo Contain
ScaleBlue Graduations
Quantity1 Pack
Graduations20 mL
AutoclavableAutoclavable
Limit of Error±20 mL
Max. Temperature80°C
Subdivision20 mL
ShapeRound
Scale ColorBlue
Base TypeHexagonal
Height (Metric)535 mm
TypeGraduated Measuring Cylinder
Unit SizeEA