Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) | Closure Size | Neck Style | Tolerance |
---|---|---|---|---|
FKS10505125 | 500 mL | 19/26 | Narrow | ±0.25 mL |
FKS10719375 | 1000 mL | 24/29 | Narrow | ±0.4 mL |
FKS10440285 | 50 mL | 14/23 | Wide | ±0.1 mL |
FKS10788805 | 100 mL | 14/23 | Narrow | ±0.1 mL |
FKS10401055 | 250 mL | 14/23 | Narrow | ±0.15 mL |