PTFE Stir Bar with Removable Ring
PTFE Stir Bar with Removable Ring
Actual product may vary
PTFE Stir Bar with Removable Ring
Provides stable stirring action. Fisherbrand™ PTFE Stir Bar with Removable Ring are autoclavable and include a pivot ring.
Have Questions?
Catalog NumberDimensions (L x W x H)
FKS10500482102 x 16 x 16mm
Catalog number FKS10500482
Price (MYR)
159.00
EA
Add to cart
Dimensions (L x W x H):
102 x 16 x 16mm
Price (MYR)
159.00
EA
Add to cart
  • Guaranteed inert and leak-free
  • Isostatically molded in pure, FDA approved, PTFE
  • Individually polished to stay cleaner for longer
Specifications
AutoclavableAutoclavable
Certifications/ComplianceCertificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
ColorWhite
DescriptionPTFE encapsulated Alnico V magnet core
Dimensions (L x W x H)102 x 16 x 16mm
For Use With (Equipment)Containers & Vessels / Stirrer Hot Plates
Height (Metric)16mm
IncludesPTFE Ring
Diameter (Metric)16 mm
Length (Metric)102 mm
Quantity1/Ea.
ShapeCylinder
Unit SizeEA