Polypropylene Straight Connectors with Conical Nozzles
Polypropylene Straight Connectors with Conical Nozzles
Actual product may vary
Polypropylene Straight Connectors with Conical Nozzles
Manufactured from polypropylene for high thermal resistance. Buerkle™ Polypropylene Straight Connectors with Conical Nozzles resist temperatures up to 135°C.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With
FKS1048689113 to 15 mm Hose
FKS106201435 to 7 mm Hose
Catalog number FKS10486891
Price (MYR)
265.00
Add to cart
For Use With:
13 to 15 mm Hose
Price (MYR)
265.00
Add to cart
Cylindrical straight hose connector with conical nozzles
  • Polypropylene
  • Autoclavable to 121°C
  • Suitable for foodstuff
  • Available in a wide range of diameters for varying hose sizes
Specifications
Certifications/ComplianceEU Foodstuffs, RoHS
AutoclavableAutoclavable
MaterialPolypropylene
For Use With13 to 15 mm Hose
Packaging Quantity10/Pk.; 10/Scale
ShapeStraight
ColorWhite
Product TypeHose Tubing Connector
Unit Size