Nalgene™ Zinc/Aluminum Hand-Operated Vacuum Pump with Gauge
Nalgene™ Zinc/Aluminum Hand-Operated Vacuum Pump with Gauge
Actual product may vary
Nalgene™ Zinc/Aluminum Hand-Operated Vacuum Pump with Gauge
Nalgene™ Zinc/Aluminum Hand-Operated Vacuum Pump with Gauge
Nalgene™ Zinc/Aluminum Hand-Operated Vacuum Pump with Gauge
Nalgene™ Zinc/Aluminum Hand-Operated Vacuum Pump with Gauge
Nalgene™ Zinc/Aluminum Hand-Operated Vacuum Pump with Gauge
Nalgene™ Zinc/Aluminum Hand-Operated Vacuum Pump with Gauge
Obtain full blank port pressure with only two strokes when using the Thermo ScientificTM NalgeneTM Zinc/Aluminum Hand-Operated Vacuum Pump with Gauge.
Have Questions?
Catalog NumberFlow RateFor Use With
FKS1048230115 cc/stroke6133-2000 Repair Kit
Catalog number FKS10482301
Price (MYR)
2,726.00
EA
Add to cart
Flow Rate:
15 cc/stroke
For Use With:
6133-2000 Repair Kit
Price (MYR)
2,726.00
EA
Add to cart
Obtain full blank port pressure with only two strokes when using the Thermo Scientific™ Nalgene™ Zinc/Aluminum Hand-Operated Vacuum Pump with Gauge, a multi-purpose self-lubricating pump featuring outstanding efficiency. Cast in corrosion-resistant zinc/aluminum and easily disassembled for repair or replacement of internal parts, this hand pump quickly and easily attains a vacuum of 25 in. (635mm) Hg or a pressure of 30 psi (35kPa).
  • Designed with ergonomic handle to make pumping easier on the hands
  • Corrosion resistant and repairable for long life in the lab
  • Pumping rate of 15cc per stroke
  • Easy-flip lever to switch from vacuum to pressure.
  • Gauge calibrated in inches of Mercury and in kPa
  • Attaches to 1/4″ (63.5cm) I.D. flexible tubing; includes a 2′ (0.7m) length of clear PVC tubing
  • Includes repair kit 6133-2000 for maintenance and repair of internal parts to extend hand pump life
Specifications
DescriptionVacuum Pump
No. per Pack1
No. per Case4
Max. Vacuum30 psig (35 kPag)
Flow Rate15 cc/stroke
TypeVacuum Pump
For Use With6133-2000 Repair Kit
Unit SizeEA