Manufactured from polypropylene for superior chemical and thermal stability. Whatman™ UNIPLATE™ Microplates are suitable for simple filtrate collection and homogeneous assay techniques used in high-throughput screening (HTS).
Have Questions?
Catalog Number
Sterility
Volume (Metric) Well
Well Shape
No. of Wells
FKS10423772
Sterile
10 mL
Round
24
Catalog number FKS10423772
Price (MYR)
4,926.00
Add to cart
Sterility:
Sterile
Volume (Metric) Well:
10 mL
Well Shape:
Round
No. of Wells:
24
Price (MYR)
4,926.00
Add to cart
Manufactured from polypropylene for superior chemical and thermal stability. Whatman™ UNIPLATE™ Microplates are suitable for simple filtrate collection and homogeneous assay techniques used in high-throughput screening (HTS).
Withstand low temperatures (-80°C) for long-term storage
Conform to ANSI™/SBS standards for compatibility with most microplate readers and automated plate-handling devices
Specifications
FormatStandard
LidWith Lid
DescriptionUNIPLATE microplates
Array4 x 6
ColorNatural
MaterialPolypropylene
Volume (Metric) Well10 mL
No. of Wells24
SterilitySterile
Well ShapeRound
For Use With (Application)Ideal for homogeneous assay techniques utilized in high-throughput screening