Blaubrand™ PURprotect™ Coated Borosilicate Glass Class A Volumetric Flask
Blaubrand™ PURprotect™ Coated Borosilicate Glass Class A Volumetric Flask
Actual product may vary
Blaubrand™ PURprotect™ Coated Borosilicate Glass Class A Volumetric Flask
Manufactured from Borosilicate Glass and complies with DIN EN ISO 1042 standards. BRAND™ Blaubrand™ PURprotect™ Coated Borosilicate Glass Class A Volumetric Flask will help to contain glass fragments in the event of breakage with its protective skin. Compared to uncoated glass flasks, the electrostatic charge is not increased. The light blue coating facilitates optical distinction.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeNeck StyleTolerance
FKS10401055250 mL14/23Narrow±0.15 mL
FKS107193751000 mL24/29Narrow±0.4 mL
FKS10505125500 mL19/26Narrow±0.25 mL
FKS1044028550 mL14/23Wide±0.1 mL
FKS10788805100 mL14/23Narrow±0.1 mL
Catalog number FKS10401055
Price (MYR)
1,027.00
PK
Add to cart
Capacity (Metric):
250 mL
Closure Size:
14/23
Neck Style:
Narrow
Tolerance:
±0.15 mL
Price (MYR)
1,027.00
PK
Add to cart
  • Manufactured from Borosilicate Glass
  • Complies with DIN EN ISO 1042 standards.
  • Calibrated to contain (TC, In).
  • The PUR coating envelops the volumetric flasks like a protective skin. In the event of breakage, the safety coating helps contain the glass fragments.
  • Compared to uncoated glass flasks the electrostatic charge is not increased.
  • The light blue coating facilitates optical distinction.
  • The maximum operating temperature at dry heat is 135°C (exposure time < 30 min).
  • Frequent autoclaving at 121 °C reduces splintering protection.
  • Cleaning up to max. 95 °C.
Specifications
CalibrationCalibrated To Contain
Certifications/ComplianceDIN EN ISO 1042
Closure MaterialPolypropylene
ColorClear with light blue coating
Capacity (Metric)250 mL
MaterialBorosilicate Glass
Neck StyleNarrow
No. per Case2 Piece
Tolerance±0.15 mL
ClassA
Closure Size14/23
Unit SizePK