Eutech™ Dissolved Oxygen Refilling Electrolyte Solution for DO Probes
Eutech™ Dissolved Oxygen Refilling Electrolyte Solution for DO Probes
Eutech™ DO meters, probes, and their accessories offer quick, accurate dissolved oxygen checks in laboratories, field and schools.
Have Questions?
Catalog Number
Capacity (Metric)
FKS10365253
60 mL
Catalog number FKS10365253
Price (MYR)
320.00
EA
Add to cart
Capacity (Metric):
60 mL
Price (MYR)
320.00
EA
Add to cart
Eutech™ Dissolved Oxygen Refilling Electrolyte Solution is used to refill Eutech™ DO probes. Eutech™ DO meters, probes, and their accessories offer quick, accurate dissolved oxygen checks in laboratories, field and schools.
Features:
60mL Bottle
Used With Probes: DO6, DO6+, DO100, DO110, DO 300, PD 300, DO 600, CD 650, PD 650, PCD 650
Specifications
Capacity (Metric)60 mL
DescriptionDO Refilling Electrolyte Solution
For Use With (Equipment)DO Probes DO6, DO6+, DO100, DO110, DO 300, PD 300, DO 600, CD 650, PD 650, PCD 650