Peristaltic Pump Tubing for Gilson™ MF1 and MF4 Medium Flow Pump Heads
Peristaltic Pump Tubing for Gilson™ MF1 and MF4 Medium Flow Pump Heads
Actual product may vary
Peristaltic Pump Tubing for Gilson™ MF1 and MF4 Medium Flow Pump Heads
Manufactured to the utmost standard. Gilson™ Peristaltic Pump Tubing for Gilson™ MF1 and MF4 Medium Flow Pump Heads are autoclavable.
Have Questions?
Catalog NumberFlow RateGas PermeabilityColorMaterial
FKS102261222mL/min.HighPink/PinkSilicone
Catalog number FKS10226122
Price (MYR)
1,493.00
PK
Add to cart
Flow Rate:
2mL/min.
Gas Permeability:
High
Color:
Pink/Pink
Material:
Silicone
Price (MYR)
1,493.00
PK
Add to cart
As part of our MINIPULS™ 3 and Evolution lines of peristaltic pumps, Gilson offers a variety of tubing of different material, inner diameter sizes, and lengths.
Choice of Materials
  • Silicone (Versilic™) tubing—is biologically neutral and does not inhibit cell culture while meeting European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993 biocompatibility.
  • ISOVERSINIC™ tubing—shows excellent resistance to strong acids, oxidizing agents, and aromatic and chlorinated solvents.
  • PharMed™ tubing—meets European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993 biocompatibility.
  • PVC (Tygon™) tubing—handles virtually all inorganic chemicals found in the lab and can be used in corrosive environments.
Specifications
AutoclavableAutoclavable
Diameter (Metric) Inner2 mm
MaterialSilicone
Overall Length (Metric)40 cm
For Use WithMF1 and MF4 medium flow pump head
For Use With (Application)Medium flow tubing has color coded retaining stops for easy recognition of the tubing's internal diameter
SterilitySterile
Flow Rate2mL/min.
Gas PermeabilityHigh
ColorPink/Pink
Product TypePeristaltic Pump Tubing
Unit SizePK