Büchner Filter Funnels with Porosity 3 VitraPOR™ Glass Filter Discs
Büchner Filter Funnels with Porosity 3 VitraPOR™ Glass Filter Discs
Actual product may vary
Büchner Filter Funnels with Porosity 3 VitraPOR™ Glass Filter Discs
Robu™ Büchner Filter Funnels with Glass Filter Discs are made of pure borosilicate 3.3 glass and are highly resistant to most chemicals, including solvents, acids, and alkalis. The VitraPOR™ filter disc is rated as Porosity 3 (Pore size: 16 to 40 μm, Standard: ISO 4793-80).
Have Questions?
Catalog NumberCapacity (Metric)Height (Metric)Diameter (Metric)TopDiameter (Metric) Stem
FKS10225551500 mL100 mm100 mm (OD), 95 mm (ID)22 mm
Catalog number FKS10225551
Price (MYR)
1,181.00
EA
Add to cart
Capacity (Metric):
500 mL
Height (Metric):
100 mm
Diameter (Metric)Top:
100 mm (OD), 95 mm (ID)
Diameter (Metric) Stem:
22 mm
Price (MYR)
1,181.00
EA
Add to cart
  • Product Type: Büchner Filter Funnel
  • Material: Borosilicate 3.3 Glass (Body), VitraPOR™ Glass (Filter)
  • Porosity: Grade 3
  • Pore Size: 16-40 μm
  • Standard: ISO 4793-80
  • For Use With (Application): Filtration
  • For Use With (Equipment): Guko Rubber Gaskets (e.g. 10430652)
Specifications
Diameter (Metric)Top100 mm (OD), 95 mm (ID)
Capacity (Metric)500 mL
Diameter (Metric) Stem22 mm
MaterialBorosilicate Glass
Height (Metric)100 mm
Certifications/ComplianceISO 4793-80
For Use With (Application)Filtration
Diameter (Metric) Disc100 mm
For Use With (Equipment)Guko Rubber Gaskets (e.g. 10430652)
PorosityGrade 3
Pore Size16 to 40 μm
Capacity (English)16.9 oz.
Product TypeBuchner Funnel
Unit SizeEA