Latex Gas Sampling Balloon
Latex Gas Sampling Balloon
Actual product may vary
Fisherbrand
Latex Gas Sampling Balloon
Gass collection bags that are constructed from latex. Kartell™ Latex Gas Sampling Balloon are best suited for collecting, containing, and carrying gas samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS102037632 L
FKS102631835 L
Catalog number FKS10203763
Price (MYR)
273.00
Add to cart
Capacity (Metric):
2 L
Price (MYR)
273.00
Add to cart
  • Cost effective
  • Ideal for collecting, containing, and carrying gas samples
Specifications
Capacity (Metric)2 L
MaterialLatex
Diameter (Metric)160 mm
Product TypeGas Sampling Balloon
Unit Size