Check Order Status
Shell Vial Kit, 8 mm, Clear Glass, PE plug
Actual product may vary
Fisherbrand
Shell Vial Kit, 8 mm, Clear Glass, PE plug
Includes 100 vials and 100 caps with pre-assembled septa.
Have Questions?
Catalog Number
Volume (Metric)
FKS10145424
1.4 mL
Catalog number
FKS10145424
Price (MYR)
191.00
Add to cart
Volume (Metric):
1.4 mL
Price (MYR)
191.00
Add to cart
ND8
Glass
Clear
Plug Cap
Star-shaped diaphragm
Polyethylene
Transparent
Reusable two compartment trays protect vials and closure while keeping matching supplies together
Specifications
Color
Clear
Diameter (Metric) Outer
8.2 mm
Material
Glass
Volume (Metric)
1.4 mL
No. per Pack
100
Height (Metric)
40 mm
Closure Size
8
Marking Spot
No
Unit Size