Check Order Status
PTFE Flat-Form Evaporating Dish
Actual product may vary
Fisherbrand
PTFE Flat-Form Evaporating Dish
Inert and chemically resistant. The PTFE flat-form evaporating dish is available in a variety of sizes.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Diameter (Metric)
Height (Metric)
FKS10088171
100 mL
80 mm
28 mm
FKS10572392
50 mL
60 mm
20 mm
2 Options
Catalog number
FKS10088171
Price (MYR)
212.00
Add to cart
Capacity (Metric):
100 mL
Diameter (Metric):
80 mm
Height (Metric):
28 mm
Price (MYR)
212.00
Add to cart
Isostatically molded in pure PTFE
Chemically resistant and inert
Super smooth internal finish
Usable to 280°C
Specifications
Capacity (English)
3.33 fl oz.
Bottom Shape
Flat
Capacity (Metric)
100 mL
Material
PTFE
Shape
Round
Type
Evaporating Dish
Interior Finish
Smooth
Temperature Range (Metric)
-200°C to +280°C
Height (Metric)
28 mm
Diameter (Metric)
80 mm
Unit Size