DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated
DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated
Actual product may vary
DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated
DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated
DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated
DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated
DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated
DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated
DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated
The gold standard for multifunctional glass laboratory bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS1004799020,000 mL
FKS11752272500 mL
FKS123471781000 mL
Catalog number FKS10047990
Price (MYR)
9,248.00
EA
Add to cart
Capacity (Metric):
20,000 mL
Price (MYR)
9,248.00
EA
Add to cart
DURAN™ Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972. The bottles are moulded from high-purity, clear, USP/EP Type I borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN™ Original GL bottles are supplied complete with iconic blue polypropylene cap and drip-free pouring ring for use up to 140°C. All components are fully autoclavable at 121 or 134°C.
  • Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
  • Retrace Code, and certificate for manufacturing lot traceability
  • Fired-on, highly durable white print
  • Suitable for long-term storage
  • Easy-to-read scale with graduation marks to estimate the content
Specifications
Capacity (Metric)20,000 mL
For Use With (Application)Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir
IncludesWith screw cap and pouring ring from PP (blue)
MaterialBorosilicate Glass (3.3)
Quantity1 Ea.
Thread Size (Metric)GL 45
AutoclavableAutoclavable
Diameter (Metric)288 mm
Unit SizeEA