Stainless Steel Test Sieve, 200 mm dia.
Stainless Steel Test Sieve, 200 mm dia.
Actual product may vary
Stainless Steel Test Sieve, 200 mm dia.
Full height test sieves provide all types of laboratory sampling and particle size analysis. Endecotts™ Stainless Steel Test Sieves, 200 mm dia. comply with ISO 3310-1 / BS410 standards.
Have Questions?
Catalog NumberPore Size
FKS1002252490 μm
Catalog number FKS10022524
Price (MYR)
1,670.00
EA
Add to cart
Pore Size:
90 μm
Price (MYR)
1,670.00
EA
Add to cart
Full height test sieves provide all types of laboratory sampling and particle size analysis. Endecotts™ Stainless Steel Test Sieves, 200 mm dia. comply with ISO 3310-1 / BS410 standards.
  • Stainless steel precision frame (ensures consistent nestability)
  • Stainless steel mesh
  • Precise aperture in accordance with ISO 3310-1 / BS410
  • Natural fillet (free flowing of sample)
  • Totally sealed (no crevice to lose material)
  • Evenly tensioned mesh (ensures accurate analysis)
  • Safe edge (big radius makes it comfortable to handle)
  • Serial number (ensures full traceability)
Specifications
Quantity1 Ea.
Pore Size90 μm
TypeTest Sieve
Certifications/ComplianceISO 3310-1 / BS410
Diameter (English) Outer7.87 in.
MaterialStainless Steel
Height (Metric)50 mm
For Use With (Application)For all types of laboratory sampling and particle size analysis
Diameter (Metric) Outer200 mm
Frame MaterialStainless Steel
Mesh MaterialStainless Steel
Unit SizeEA