Nalgene™ Tubing Y-Type Connectors
Nalgene™ Tubing Y-Type Connectors
Actual product may vary
Nalgene™ Tubing Y-Type Connectors
Maximize liquid flow with non-tapered hose barbed Y-Type Connectors, molded of polypropylene resin, free of animal derived components.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With
FKS100206015/16 in. I.D.
FIS15-320-10B3/16 in. I.D.
Catalog number FKS10020601
Price (MYR)
449.00
PK
Add to cart
For Use With:
5/16 in. I.D.
Price (MYR)
449.00
PK
Add to cart
Maximize liquid flow with non-tapered hose barbed Thermo Scientific™ Nalgene™ Tubing Y-Type Connectors, molded of polypropylene resin, free of animal derived components. Products are double-bagged to maintain cleanliness.
  • Polypropylene construction
  • Autoclavable
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionY-Type Connector
AutoclavableAutoclavable
Diameter (English) Inner5/16 in.
Diameter (Metric) Inner7.93 mm
MaterialPolypropylene
For Use With5/16 in. I.D.
Packaging Quantity12/Pk., 72/Cs.
Product TypeTubing Connector
Unit SizePK