HDPE, Jerry Can
HDPE, Jerry Can
Actual product may vary
HDPE, Jerry Can
Canister for storage or transport. Canister with UN transport approval and tamper-evident seal.
Have Questions?
Catalog NumberCapacity (Metric)Dimensions (L x W x H)
FKS1000679110 L230 x 190 x 310 mm
Catalog number FKS10006791
Price (MYR)
136.00
EA
Add to cart
Capacity (Metric):
10 L
Dimensions (L x W x H):
230 x 190 x 310 mm
Price (MYR)
136.00
EA
Add to cart
  • For storage or transport
  • HDPE
  • With UN approval for transport
  • Screw lid without drain valve is included, screw lid with drain valve available as accessory
Specifications
AutoclavableNot Autoclavable
Capacity (English)2.6 gal.
Capacity (Metric)10 L
Certifications/ComplianceUN approval 3H1/X1.9/250
Closure Size1427-9105, Orig. Screw Cap DIN50
Closure TypeDIN 50
ColorTransparent
Diameter (Metric) Inner Neck40.5 mm
Dimensions (L x W x H)230 x 190 x 310 mm
Dimensions (L x W)230 x 190 mm
For Use With (Application)For Storage or Transport
Height (Metric) With Closure310 mm
IncludesScrew lid
Length (Metric)230 mm
MaterialHDPE
Mouth StyleS50 Thread
Product TypeJerry Can
ShapeSquare
SterilityNon-sterile
Width (Metric)190 mm
Unit SizeEA