HyClone™ Phosphate Buffered Saline (PBS)
HyClone™ Phosphate Buffered Saline (PBS)
Actual product may vary
HyClone™ Phosphate Buffered Saline (PBS)
Buffer cell culture growth with PBS
Have Questions?
Catalog NumberQuantityConcentration
FISSH30256021000 mL1 X
Catalog number FISSH3025602
Price (MYR)
-
Quantity:
1000 mL
Concentration:
1 X
  • QC-tested solution
Specifications
Certifications/CompliancecGMP and ISO 9001:2000 Guidelines
Concentration1 X
FormLiquid
Product TypePhosphate Buffered Saline (PBS)
Sterilization Method0.1 μm Sterile-filtered
Without AdditivesNo Calcium, No Magnesium, No Phenol Red
Endotoxin Level=1.0EU/mL
pH7.0 to 7.2
Quantity1000 mL
Unit SizeEA
Contents & Storage
15° to 30°C