Featherweight Entomology Forceps, Spring Steel
Featherweight Entomology Forceps, Spring Steel
Actual product may vary
Fisherbrand
Featherweight Entomology Forceps, Spring Steel
Featherweight Entomology Forceps can be bent in virtually any direction providing optimal control and ease if use. Ideal for handling larvae or eggs without crushing them or specimens stored in vials.
Have Questions?
Catalog NumberTip Style
FISS72114Broad Point
Catalog number FISS72114
Price (MYR)
11.75
Add to cart
Tip Style:
Broad Point
Price (MYR)
11.75
Add to cart
  • These forceps are crafted from flexible steel, allowing them to be bent in virtually any direction, providing optimal control and maneuverability during use.
  • With a length of 4.5 inches, they are compact and easy to handle, making them ideal for working in tight spaces.
Specifications
Product TypeEntomology Forceps
DescriptionFlexible Forceps
MaterialSpring Steel
Tip StyleBroad Point
Length (English)4.5 in.
Length (Metric)10.8 cm
Unit Size