Chemically-Resistant Wire Racks
Chemically-Resistant Wire Racks
Actual product may vary
Chemically-Resistant Wire Racks
Chemically-Resistant Wire Racks
Chemically-Resistant Wire Racks
Chemically-Resistant Wire Racks
HDPE coated wire racks are durable for transport or sample handling. Chemically-Resistant Wire Racks suitable for use with conical, round, or flat-bottom plastic tubes and bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPositionsDiameter (Metric) Well
FISS68125FB4016 to 20 mm
FISS68126FB4813 to 16 mm
FISS43521FB7210 to 13 mm
Catalog number FISS68125FB
Price (MYR)
166.00
EA
Add to cart
Positions:
40
Diameter (Metric) Well:
16 to 20 mm
Price (MYR)
166.00
EA
Add to cart
  • HDPE Coated Wire Racks
  • Chemically-resistant
  • Holds all brands and types of conical, round or flat-bottom, glass or plastic tubes and/or bottles
  • Square wells and three deck levels keep tubes secure
  • Use between temperatures of -40°C and 121°C
  • Slotted Bottom Support
  • Autoclavable
Specifications
Product TypeTube Rack
DescriptionFisherbrand HDPE Coated Wire Rack
Array4 x 10
AutoclavableAutoclavable
ColorBlue
HoldsTubes
MaterialHDPE-coated Wire
Positions40
StackableNo
SterilityNon-sterile
For Use With (Application)Holds Tubes
No. per Case48/Cs.
No. per Pack1 Ea.
Temperature (Metric) Range-40°C and 121°C
Dimensions (L x W x H)9.50 x 3.90 x 3.10 in.
Length (Metric)24.13 cm
Length (English)9.50 in.
Width (Metric)9.91 cm
Width (English)3.90 in.
Height (Metric)7.87 cm
Height (English)3.10 in.
Diameter (Metric) Well16 to 20 mm
Weight (English)0.4 lb.
Weight (Metric)0.18 kg
Unit SizeEA