Check Order Status
Glass Funnel Filters
Actual product may vary
Glass Funnel Filters
Eisco™ Glass Funnel Filters are made from high quality borosilicate glass and designed with an angled stem and beaded rim for durability.
Have Questions?
Change view
Catalog Number
Diameter (Metric)Top
Diameter (Metric) Stem
Length (Metric) Stem
FISS41650
150 mm
16 mm
15 cm
FISS89478
125 mm
16 mm
11.43 cm
2 Options
Catalog number
FISS41650
Price (MYR)
157.00
EA
Add to cart
Diameter (Metric)Top:
150 mm
Diameter (Metric) Stem:
16 mm
Length (Metric) Stem:
15 cm
Price (MYR)
157.00
EA
Add to cart
Funnel filters made from high quality borosilicate glass
60° angle at bottom of stem
Autoclavable
Specifications
Diameter (Metric)Top
150 mm
Capacity (Metric)
1000 mL
Diameter (Metric) Stem
16 mm
Material
Borosilicate Glass
Height (Metric)
26 cm
Angle
60°
Autoclavable
Autoclavable
Color
Clear
Cone Size
15 cm
Graduated
No
For Use With (Application)
Filtering, Pouring
Length (Metric) Stem
15 cm
Quantity
1 Ea.
Wall Thickness
1.5 mm
Product Type
Filter Funnel