Glass Erlenmeyer Flasks with PTFE Lined Screw Cap
Glass Erlenmeyer Flasks with PTFE Lined Screw Cap
Actual product may vary
Fisherbrand
Glass Erlenmeyer Flasks with PTFE Lined Screw Cap
Erlenmeyer flasks made from high quality borosilicate glass. Includes an acid resistant polypropylene screw cap that is lined with PTFE. Both flask and cap are autoclavable.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FISS40695250 mL
FISS40696500 mL
Catalog number FISS40695
Price (MYR)
29.39
Add to cart
Capacity (Metric):
250 mL
Price (MYR)
29.39
Add to cart
  • Erlenmeyer conical shaped flask
  • Made from high quality borosilicate glass
  • Includes PTFE lined, acid resistant polypropylene screw cap
  • Flask and cap are both autoclavable
Specifications
Closure ColorBlue
Closure MaterialPolypropylene, PTFE
ColorClear
Capacity (Metric)250 mL
MaterialBorosilicate Glass (3.3)
For Use With (Application)Store liquids, culturing
Graduation Range100 to 250 mL
Graduations50 mL
Height (Metric)12.7 cm
IncludesFlask, Screw Cap
Length (Metric)7 cm
Product TypeErlenmeyer Flasks
Width (Metric)7 cm
AutoclavableAutoclavable
DisposableReusable
GraduatedYes
Mouth StyleNarrow
SterilityNon-sterile
Bottom ShapeFlat
Closure TypeScrew Cap
Surface TreatmentNon-treated
Unit Size