Glass Eudiometer Tube with Platinum Electrodes
Glass Eudiometer Tube with Platinum Electrodes
Actual product may vary
Fisherbrand
Glass Eudiometer Tube with Platinum Electrodes
Eudiometer Tubes are closed at one end and include two sealed platinum electrodes. Ideal for measuring the volume gas produced or consumed in a chemical reaction.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FISS2734650 mL
FISS89304100 mL
Catalog number FISS27346
Price (MYR)
80.44
Add to cart
Capacity (Metric):
50 mL
Price (MYR)
80.44
Add to cart
  • For measuring the volume of gas produced or consumed in a chemical reaction.
  • Constructed of high quality borosilicate glass.
  • Two platinum electrodes are sealed.
  • Tube is closed at one end.
  • White, 0.20 mL graduations.
Specifications
Product TypeEudiometer Tube
ColorClear
DescriptionPlatinum electrodes
Grade9 to 12
For Use With (Application)Measuring the volume of gas produced or consumed in a chemical reaction
Includes2 Platinum Elecrodes
MaterialBorosilicate Glass
Quantity1 Ea.
Weight (English)0.4 lb.
Weight (Metric)181.43 g
Length (English)27 in.
Length (Metric)685.8 mm
Capacity (Metric)50 mL
Class Size1 Lab Group
Unit Size