Round and Tapered Micro Spatula
Round and Tapered Micro Spatula
Actual product may vary
Round and Tapered Micro Spatula
Made from high quality stainless steel, spatula has one rounded end and one tapered end
Have Questions?
Catalog number FISS27087
Price (MYR)
31.00
Add to cart
Price (MYR)
31.00
Add to cart
  • Round and Tapered Micro Spatula
  • Made from high quality stainless steel
  • One rounded end, one tapered end
  • Overall Length: 8 in.
  • Wire Diameter: 4 mm
  • Great for any school or home laboratory
Specifications
Quantity1
AutoclavableAutoclavable
ShapeSpoon End, Spatula End
Product TypeDouble Ended Micro Spatula
Length (English)8 in.
Diameter (Metric) Outer4 mm
MaterialStainless Steel
Length (Metric)20 cm
Unit Size