Semi Micro Test Tube Brush
Semi Micro Test Tube Brush
Actual product may vary
Semi Micro Test Tube Brush
Semi Micro Test Tube Brush for thoroughly washing 6 to 10 mm test tubes.
Have Questions?
Catalog NumberColor
FISS27023White
Catalog number FISS27023
Price (MYR)
3.54
Add to cart
Color:
White
Price (MYR)
3.54
Add to cart
  • Nylon Semi-Micro Test Tube Cleaning Brush
  • Twisted wire handle
  • Fiber Section is 2.75 in. long with a 10 mm diameter
  • Ideal for semi-micro test tubes with diameters 6 to 10 mm
  • Overall Length: 9 in.
Specifications
Product TypeCleaning Brush
Bristle MaterialNylon
Bristle TypeFlexible
Color-codedNo
Handle MaterialStainless Steel Wire
Handle TypeTwisted Wire
Tip TypeFan Tip
ScentUnscented
SterilityNon-sterile
ColorWhite
Diameter (Metric) Bristle10 mm
For Use With (Equipment)Test Tubes
Length (English)9 in.
Length (English) Bristle2.75 in.
Length (Metric)23 cm
Size6 to 10 mm
Unit Size