Check Order Status
pH Paper Strips
Actual product may vary
pH Paper Strips
An easy choice for classroom experiments or demonstrations
Have Questions?
Change view
Catalog Number
Type
pH Range
FISS22508
pH Test Paper
0 to 14
FISS22512
pH Test Paper
8 to 9.5
2 Options
Catalog number
FISS22508
Price (MYR)
-
Type:
pH Test Paper
pH Range:
0 to 14
Ideal for educational use
Low-cost paper strips are easy to pass around the classroom
Easy-to-usedash (em)simply dip strip in solution to be tested and match to color chart immediately
Specifications
pH Range
0 to 14
For Use With (Application)
For classroom experiments or demonstrations
Graduations
1
Quantity
100 Tests/Vial, 10 Vials/Cs.
Type
pH Test Paper