Kimble™ BioMasherII™ Closed System Micro Tissue Homogenizer
Kimble™ BioMasherII™ Closed System Micro Tissue Homogenizer
Actual product may vary
DWK Life Sciences
Kimble™ BioMasherII™ Closed System Micro Tissue Homogenizer
Ensure efficient grinding of samples by minimizing potential exposure to hazardous materials
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPackagingSterility
FISK7496250010BulkNon-sterile
FISK7496250030Single packSterile
Catalog number FISK7496250010
Price (MYR)
1,363.00
CS
Add to cart
Packaging:
Bulk
Sterility:
Non-sterile
Price (MYR)
1,363.00
CS
Add to cart
Ensure efficient grinding of samples by minimizing potential exposure to hazardous materials
  • Clear polypropylene tube
  • Polyacetal pestle with molded-in shaft guard
  • Abrasive surface (pestle tip and inner tube area)
  • Available in Sterile and non-sterile versions
  • Capacity: 1.5mL
For animal tissues and organs, plant materials, insects, DNA, RNA, proteins, yeasts and enzymes
Specifications
Capacity (English)0.05 oz.
For Use With (Application)For animal tissues and organs, plant materials, insects, DNA, RNA, Proteins, yeasts and Enzymes
AutoclavableAutoclavable
ColorClear
IncludesPP tube, POM pestle
Capacity (Metric)1.5 mL
PackagingBulk
Capacity0.05 oz., 1.5mL