Check Order Status
Mortar and Pestle Regular Form
Actual product may vary
Mortar and Pestle Regular Form
Mortar and Pestle Regular Form
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Height (Metric)
FISFB970A
1.5 mL
19mm
FISFB970C
9 mL
28mm
2 Options
Catalog number
FISFB970A
Price (MYR)
359.00
EA
Add to cart
Capacity (Metric):
1.5 mL
Height (Metric):
19mm
Price (MYR)
359.00
EA
Add to cart
Low permeability
High strength
Hard
High resistance to chemical attacks
Translucent or glazed body
Specifications
Capacity (Metric)
1.5 mL
Diameter (Metric) Inner
20mm
Diameter (Metric) Outer
25mm
Product Type
Mortar and Pestle
Material
Agate Stone
For Use With (Application)
For grinding or crushing
Height (Metric)
19mm
Includes
Mortar and Pestle