Check Order Status
Porcelain Buchner Funnels with Fixed Perforated Plates
Actual product may vary
Fisherbrand
Porcelain Buchner Funnels with Fixed Perforated Plates
For suction filtration applications
Have Questions?
Catalog Number
Capacity (Metric)
Diameter (Metric) Disc
Diameter (Metric) Plate
FISFB966J
1060 mL
142 - 150mm
151mm
Catalog number
FISFB966J
Price (MYR)
259.00
Add to cart
Capacity (Metric):
1060 mL
Diameter (Metric) Disc:
142 - 150mm
Diameter (Metric) Plate:
151mm
Price (MYR)
259.00
Add to cart
For suction filtration applications
Autoclavable
Highly resistant to chemical attack
Without cover
Glazed inside and outside with the exception of the rim
Specifications
Diameter (Metric)Top
17cm
Capacity (Metric)
1060 mL
Material
Porcelain
Height (Metric)
22.1 cm
Autoclavable
Autoclavable
Diameter (Metric) Plate
151mm
For Use With (Application)
Suction filtration applications
Diameter (Metric) Disc
142 - 150mm
Includes
Fixed perforated plate
Quantity
Each
Wall Thickness
5.8mm
Weight (Metric)
1.28kg
Product Type
Buchner Funnel
Unit Size