Porcelain Buchner Funnels with Fixed Perforated Plates
Porcelain Buchner Funnels with Fixed Perforated Plates
Actual product may vary
Fisherbrand
Porcelain Buchner Funnels with Fixed Perforated Plates
For suction filtration applications
Have Questions?
Catalog NumberCapacity (Metric)Diameter (Metric) DiscDiameter (Metric) Plate
FISFB966H700 mL110 - 124mm126mm
Catalog number FISFB966H
Price (MYR)
190.00
Add to cart
Capacity (Metric):
700 mL
Diameter (Metric) Disc:
110 - 124mm
Diameter (Metric) Plate:
126mm
Price (MYR)
190.00
Add to cart
For suction filtration applications
  • Autoclavable
  • Highly resistant to chemical attack
  • Without cover
  • Glazed inside and outside with the exception of the rim
Specifications
Diameter (Metric)Top14.2cm
Capacity (Metric)700 mL
MaterialPorcelain
Height (Metric)20.2 cm
AutoclavableAutoclavable
Diameter (Metric) Plate126mm
For Use With (Application)Suction filtration applications
Diameter (Metric) Disc110 - 124mm
IncludesFixed perforated plate
QuantityEach
Wall Thickness5.5mm
Weight (Metric)0.9kg
Product TypeBuchner Funnel
Unit Size