Porcelain Buchner Funnels with Fixed Perforated Plates
Porcelain Buchner Funnels with Fixed Perforated Plates
Actual product may vary
Fisherbrand
Porcelain Buchner Funnels with Fixed Perforated Plates
For suction filtration applications
Have Questions?
Catalog NumberCapacity (Metric)Diameter (Metric) DiscDiameter (Metric) Plate
FISFB966G550 mL110mm114mm
Catalog number FISFB966G
Price (MYR)
140.00
Add to cart
Capacity (Metric):
550 mL
Diameter (Metric) Disc:
110mm
Diameter (Metric) Plate:
114mm
Price (MYR)
140.00
Add to cart
For suction filtration applications
  • Autoclavable
  • Highly resistant to chemical attack
  • Without cover
  • Glazed inside and outside with the exception of the rim
Specifications
Diameter (Metric)Top12.5cm
Capacity (Metric)550 mL
MaterialPorcelain
Height (Metric)19.5 cm
AutoclavableAutoclavable
Diameter (Metric) Plate114mm
For Use With (Application)Suction filtration applications
Diameter (Metric) Disc110mm
IncludesFixed perforated plate
QuantityEach
Wall Thickness5mm
Weight (Metric)0.66kg
Product TypeBuchner Funnel
Unit Size