Porcelain Crucible Covers
Porcelain Crucible Covers
Actual product may vary
Fisherbrand
Porcelain Crucible Covers
For use with Fisherbrand High- and Wide-Form Porcelain Crucibles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHeight (Metric)Thickness (Metric)Diameter (Metric) OuterDiameter (Metric) Inner
FISFB965T6mm1.9mm44mm38mm
FISFB965V7mm2.5mm60mm54mm
FISFB965Z9mm3mm115mm105mm
Catalog number FISFB965T
Price (MYR)
101.00
Add to cart
Height (Metric):
6mm
Thickness (Metric):
1.9mm
Diameter (Metric) Outer:
44mm
Diameter (Metric) Inner:
38mm
Price (MYR)
101.00
Add to cart
For use with Fisherbrand High- and Wide-Form Porcelain Crucibles
  • Glazed inside and out except for the outside bottom surface
  • With perforated handle
  • Strongly recommended that heating and cooling rates not exceed 200°C per hour and maximum use temperature of 1100°C
Specifications
DescriptionWith Perforated handle
Diameter (Metric) Inner38mm
MaterialPorcelain
FinishGlazed
For Use With (Application)Use with Fisherbrand high and wide form porcelain crucible
Max. Temperature (Metric)1150°C
Thickness (Metric)1.9mm
Weight (Metric)14g
Diameter (Metric) Outer44mm
Height (Metric)6mm
Product TypeCrucible Cover
Unit Size