Wide-Form Porcelain Crucibles
Wide-Form Porcelain Crucibles
Actual product may vary
Fisherbrand
Wide-Form Porcelain Crucibles
Withstand temperatures up to 1150°C (2102°F)
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (Metric)Diameter (Metric) OuterWeight (Metric)
FISFB965N150 mL52mm84mm90g
FISFB965K30 mL31mm50mm26g
FKS1342971930 mL31mm50mm26g
FISFB965L50 mL37mm61mm35g
FISFB965P250 mL60mm102mm124g
Catalog number FISFB965N
Price (MYR)
129.00
Add to cart
Capacity (Metric):
150 mL
Height (Metric):
52mm
Diameter (Metric) Outer:
84mm
Weight (Metric):
90g
Price (MYR)
129.00
Add to cart
Withstand temperatures up to 1150°C (2102°F)
  • Glazed inside and out except for outside bottom surface and rim
Specifications
MaterialPorcelain
FinishGlazed
Max. Temperature (Metric)1150°C
Thickness (Metric)2.3mm
Weight (Metric)90g
Capacity (Metric)150 mL
Diameter (Metric) Outer84mm
Height (Metric)52mm
Product TypeCrucible
Unit Size