Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) | Height (Metric) | Thickness (Metric) | Diameter (Metric) Outer |
---|---|---|---|---|
FISFB965F | 40 mL | 40mm | 2 mm | 50mm |
FISFB965E | 30 mL | 37mm | 2mm | 43mm |
FKS13449699 | 30 mL | 37mm | 2mm | 43mm |
FISFB965G | 50 mL | 43mm | 2.1mm | 53mm |
FKS13489699 | 50 mL | 43mm | 2.1mm | 53mm |
FKS13499699 | 100 mL | 54mm | 2.1mm | 65mm |
FKS13409709 | 40 mL | 40mm | 2 mm | 50mm |