Porcelain Mortars
Porcelain Mortars
Actual product may vary
Fisherbrand
Porcelain Mortars
Glazed outside except bottom and unglazed inside
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityDiameter (English) OuterDiameter (Metric) Outer
FISFB961C5.07 oz., 145mL3.54 in.9cm
FISFB961E13.5 oz., 400mL5.11 in.13cm
Catalog number FISFB961C
Price (MYR)
50.00
Add to cart
Capacity:
5.07 oz., 145mL
Diameter (English) Outer:
3.54 in.
Diameter (Metric) Outer:
9cm
Price (MYR)
50.00
Add to cart
Glazed outside except bottom and unglazed inside
  • With lip
  • Deep form
  • Export quality
  • Color: white
  • Available in various capacities
Specifications
Capacity (Metric)145 mL
FinishGlazed Outer
Capacity (English)5.07 oz.
Diameter (English) Inner2.95 in.
Diameter (English) Outer3.54 in.
Diameter (Metric) Inner7.5cm
Diameter (Metric) Outer9cm
Product TypeMortar
MaterialPorcelain
Height (English)2.75 in.
Height (Metric)7cm
Capacity5.07 oz., 145mL
Unit Size