Check Order Status
Reusable Glass Short-Stem Funnels
Actual product may vary
Fisherbrand
Reusable Glass Short-Stem Funnels
High resistance to chemical attack, mechanical, and thermal shock
Have Questions?
Change view
Catalog Number
Capacity (Metric)
FISFB60158100
225 mL
FISFB-601-5875
90 mL
FISFB6015865
60 mL
3 Options
Catalog number
FISFB60158100
Price (MYR)
714.00
Add to cart
Capacity (Metric):
225 mL
Price (MYR)
714.00
Add to cart
High resistance to chemical attack, mechanical, and thermal shock
Constructed with heavy uniform walls, molded rims, and fire-polished stems
Formed to a 58° angle for faster, more effective filtering
Cone is suspended by its uppermost edge, leaving most of the conical area for filtration
Specifications
Diameter (Metric)Top
100mm
Capacity (Metric)
225 mL
Diameter (Metric) Stem
9mm
Material
Borosilicate Glass
Certifications/Compliance
ASTM E1095, Type I, Class A
Angle
58°
Diameter (Metric) Plate
100mm
Diameter (Metric) Filter
185mm
Length (Metric) Stem
97mm
Unit Size