Reusable Glass Narrow-Mouth Erlenmeyer Flasks
Reusable Glass Narrow-Mouth Erlenmeyer Flasks
Actual product may vary
Reusable Glass Narrow-Mouth Erlenmeyer Flasks
Reusable Glass Narrow-Mouth Erlenmeyer Flasks
Fisherbrand
Reusable Glass Narrow-Mouth Erlenmeyer Flasks
Long, tapered outside contour minimizes chipping
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FISFB500500500 mL
FISFB500125125 mL
FISFB50010001000 mL
FKS154461431000 mL
FKS15436143500 mL
FISFB500250250 mL
Catalog number FISFB500500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
500 mL
Fisherbrand™ Reusable Glass Narrow-Mouth Erlenmeyer Flasks are useful for measuring and mixing solutions where a high degree of accuracy is not necessary. Long, tapered outside contour minimizes chipping.
  • Reinforced top tooled with a rounded finish
  • Thick-walled body
  • White ceramic enamel marking spot
Specifications
Closure MaterialRubber
DescriptionTop style: Heavy duty tooled with capacity scale
Diameter (English) Outer4.01 in.
Capacity (Metric)500 mL
MaterialBorosilicate Glass
Graduation Range100 to 500 mL
Graduations50 mL
Height (English)6.85 in.
Height (Metric)17.4 cm
No. of Positions1
Writing SurfaceMatte Finish Ceramic Enamel Marking
DisposableReusable
Mouth StyleNarrow
Diameter (Metric) Outer102 mm
Closure Size7
Unit Size