GHS Labeled Right-to-Know Safety-Vented Wash Bottles
GHS Labeled Right-to-Know Safety-Vented Wash Bottles
Actual product may vary
GHS Labeled Right-to-Know Safety-Vented Wash Bottles
GHS Labeled Right-to-Know Safety-Vented Wash Bottles
GHS Labeled Right-to-Know Safety-Vented Wash Bottles
Fisherbrand
GHS Labeled Right-to-Know Safety-Vented Wash Bottles
Unique valve prevents pressure build-up, eliminating potentially dangerous chemical drips and allows dispensing of wash bottle in upright or inverted position.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberChemical Label
FISFB11865155Isopropanol
FISFB11865150Acetone
FISFB11865163Ethanol
FISFB11865160Sodium Hypochlorite
Catalog number FISFB11865155
Price (MYR)
294.00
Add to cart
Chemical Label:
Isopropanol
Price (MYR)
294.00
Add to cart
  • Pre-printed bottles with chemical name and formula, GHS pictogram and signal word; National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
  • Translucent LDPE bottles, Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm
  • Wide mouth 53mm (2 in.) for easy refilling
  • Color-coded polypropylene closures aid in quick and easy identification
  • Assortment bag includes one each: Acetone, Isopropanol, Bleach and Ethanol
    Bottle with Label: TOLUENE
    • Red LDPE bottle
    • Does not include vented valve
    Bottle with Label: DEIONIZED H2O, DISTILLED H20 and WATER
    • Does not indicate hazardous chemical and does not have/require GHS pictogram or signal word
    Bottle with Label: METHYL ETHYL KETONE
    • Does not include vented valve
    Compliance: Globally Harmonized System of Classification and Labeling of Chemicals (GHS) and OSHA’s HazCom 2012 requirements – as pertains to “Workplace Labels”.
Specifications
Capacity (Metric)500 mL
ColorNatural
MaterialLDPE
Quantity3/Bag
Closure ColorYellow
AutoclavableNot Autoclavable
MouthWide
Chemical LabelIsopropanol
Closure MaterialPolypropylene
Closure Size53 mm
Closure TypeCap
Unit Size