Solvent-Vented™ and Safety-Labeled Wash Bottles
Solvent-Vented™ and Safety-Labeled Wash Bottles
Actual product may vary
Solvent-Vented™ and Safety-Labeled Wash Bottles
Solvent-Vented™ and Safety-Labeled Wash Bottles
Solvent-Vented™ and Safety-Labeled Wash Bottles
Solvent-Vented™ and Safety-Labeled Wash Bottles
Pre-printed with Chemical name and formula. Fisherbrand™ Solvent-Vented™ and Safety-Labeled Wash Bottles have a patented valve which eliminates drips and allows bottle to dispense upright or inverted.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberChemical Label
FISFB0340934Unlabeled
FISFB0340937Acetone
FISFB0340936Ethanol
FISFB0340935Isopropanol
Catalog number FISFB0340934
Price (MYR)
248.00
PK
Add to cart
Chemical Label:
Unlabeled
Price (MYR)
248.00
PK
Add to cart
  • Wide mouth 2 in. (53mm) bottle for easy refilling
  • Polypropylene closure with leakproof design, color-coded to aid in easy identification
  • Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
  • Labeled with six of the most commonly used solvent names or label your own; custom labeling available
Use LYOB only with solvents compatible with LDPE.
Specifications
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorTranslucent
MaterialLDPE
Closure ColorNatural
Certifications/ComplianceOSHA 29 CFR 1910.1200
AutoclavableNot Autoclavable
MouthWide
Chemical LabelUnlabeled
Closure MaterialPolypropylene
Closure Size53 mm
Unit SizePK