Translucent White Wash Bottles
Translucent White Wash Bottles
Actual product may vary
Translucent White Wash Bottles
Translucent White Wash Bottles
Translucent White Wash Bottles
Fisherbrand
Translucent White Wash Bottles
Versatile bottles with single drop or fine stream options.
Have Questions?
Catalog NumberCapacity (Metric)
FISFB0340922C500 mL
Catalog number FISFB0340922C
Price (MYR)
390.00
Add to cart
Capacity (Metric):
500 mL
Price (MYR)
390.00
Add to cart
  • LDPE with one-piece polypropylene screw cap and nozzle
  • Tip may be cut back to increase flow
Specifications
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorTranslucent
MaterialLDPE
No. per Pack6
AutoclavableNot Autoclavable
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeScrew Cap
Unit Size