Amber Boston Round Glass Bottles with Black Phenolic Caps
Amber Boston Round Glass Bottles with Black Phenolic Caps
Actual product may vary
Amber Boston Round Glass Bottles with Black Phenolic Caps
Packaged in convenience packs with black phenolic caps attached to maintain cleanliness. Fisherbrand™ Amber Boston Round Glass Bottles with Black Phenolic Caps are ideal for solvent, chemical or sample storage.
Have Questions?
Catalog NumberCapacity (Metric)
FISFB02911899950 mL
Catalog number FISFB02911899
Price (MYR)
573.00
CS
Add to cart
Capacity (Metric):
950 mL
Price (MYR)
573.00
CS
Add to cart
  • Glass offers minimal chemical interference with contents
  • Amber glass bottles absorb all wavelengths in the ultraviolet spectrum
  • Black Phenolic Caps form an exceptionally tight seal and offer a good chemical barrier
  • Liners molds themselves around the sealing areas of the finish as the cap is screwed on, virtually eliminating leakage, evaporation, contamination, binding and backing off
  • Ideal for everything from liquids, to soaps, to macro-invertebrates
Specifications
Capacity (Metric)950 mL
Capacity (English)32 oz.
ColorAmber
Diameter (Metric) Outer Neck29 mm
Diameter (Metric) Inner Neck20 mm
Height (Metric) With Closure209 mm
MaterialGlass (Type 3)
Quantity30
Closure ColorBlack
Neck Finish33-400
Closure IncludedYes
MouthNarrow
Height (Metric)206 mm
Product TypeBoston Round Bottle
Closure LinerPolyethylene Cone
Closure MaterialPhenolic
Closure Size33-400 mm
Diameter (Metric)94 mm
Unit SizeCS