Clear Standard Wide Mouth Glass Bottles with White Polypropylene Caps
Clear Standard Wide Mouth Glass Bottles with White Polypropylene Caps
Actual product may vary
Clear Standard Wide Mouth Glass Bottles with White Polypropylene Caps
Clear, Type III soda-lime glass ideal for general, small to large volume storage
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Liner
FISFB02911731120 mLPulp/Vinyl
FISFB02911916240 mLPulp/Vinyl
FKS16154437240 mLPTFE/Polyethylene Foam
FISFB02911917480 mLPulp/Vinyl
Catalog number FISFB02911731
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
120 mL
Closure Liner:
Pulp/Vinyl
  • Features a wide mouth opening for easy transfer of contents
  • Applications include pharmaceutical, chemical, environmental, and food packaging and storing
  • Manufactured from clear flint glass
Specifications
Capacity (Metric)120 mL
Capacity (English)4 oz.
ColorClear
Diameter (Metric) Outer Neck45 mm
Diameter (Metric) Inner Neck35 mm
Height (Metric) With Closure89 mm
MaterialGlass (Type 3)
Quantity24
Closure ColorWhite
Neck Finish48-400
Closure IncludedYes
MouthWide
Height (Metric)86 mm
Product TypeStandard Wide Mouth Bottle
Closure LinerPulp/Vinyl
Closure MaterialPolypropylene
Closure Size48-400 mm
Diameter (Metric)51 mm
Unit Size