Clear Graduated Glass Bottles with Black Phenolic Caps
Clear Graduated Glass Bottles with Black Phenolic Caps
Actual product may vary
Clear Graduated Glass Bottles with Black Phenolic Caps
Fisherbrand™ Clear Graduated Glass Bottles with Black Phenolic Caps feature a wide mouth for easy removal of contents.
Have Questions?
Catalog NumberCapacity (Metric)
FISFB02911464B60 mL
Catalog number FISFB02911464B
Price (MYR)
414.00
CS
Add to cart
Capacity (Metric):
60 mL
Price (MYR)
414.00
CS
Add to cart
  • Graduated in both milliliters and ounces
  • Clear glass bottles are designed with a slight shoulder for easy pouring and handling
  • Wide mouth facilitates easy mixing, handling, and transfer of contents
Specifications
Capacity (Metric)60 mL
Capacity (English)2 oz.
ColorClear
MaterialGlass (Type 3)
Quantity48
Closure ColorBlack
Neck Finish38-400
Closure IncludedYes
GraduatedYes
MouthWide
Closure LinerPolyethylene Cone
Closure MaterialPhenolic
Unit SizeCS