Clear Graduated Glass Bottles with White Polypropylene Caps
Clear Graduated Glass Bottles with White Polypropylene Caps
Actual product may vary
Clear Graduated Glass Bottles with White Polypropylene Caps
Fisherbrand™ Clear Graduated Glass Bottles with White Polypropylene Caps feature a wide mouth for easy removal of contents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Quantity
FISFB02911453A30 mL48
FKS16154377240 mL24
Catalog number FISFB02911453A
Price (MYR)
210.00
Add to cart
Capacity (Metric):
30 mL
Quantity:
48
Price (MYR)
210.00
Add to cart
  • Graduated in both milliliters and ounces
  • Clear glass bottles are designed with a slight shoulder for easy pouring and handling
  • Wide mouth facilitates easy mixing, handling, and transfer of contents
Specifications
Capacity (Metric)30 mL
Capacity (English)1 oz.
ColorClear
Diameter (Metric) Outer Neck29 mm
Diameter (Metric) Inner Neck20 mm
Height (Metric) With Closure71 mm
MaterialGlass (Type 3)
Quantity48
Closure ColorWhite
Neck Finish33-400
Closure IncludedYes
GraduatedYes
MouthWide
Height (Metric)68 mm
Product TypeGraduated Bottle
Closure LinerPulp/Vinyl
Closure MaterialPolypropylene
Closure Size33-400 mm
Diameter (Metric)34 mm
Unit Size