Check Order Status
Amber Wide Mouth Packer Glass Bottles with White Polypropylene Caps
Actual product may vary
Fisher Catalogue
Amber Wide Mouth Packer Glass Bottles with White Polypropylene Caps
Fisherbrand™ Amber Wide Mouth Packer Glass Bottles with White Polypropylene Caps are ideal for storage of light sensitive samples or dry powders.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Quantity
FISFB02911434A
120 mL
24
FISFB02911432A
30 mL
24
FISFB02911433A
60 mL
24
FISFB02911436A
500 mL
12
4 Options
Catalog number
FISFB02911434A
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
120 mL
Quantity:
24
Feature a wide opening for making them the ideal general purpose bottle for solids
The amber color filters out ultraviolet wavelengths, making amber packers ideal for light sensitive solids
Amber packers are the classic vitamin bottles and are often used in pharmaceutical packaging
Easy transfer of contents
Good impact strength and pliability
Caps can be remelted after they are formed
Liner is made of a vinyl coating applied to high density polyethylene coated paper and then it is laminated to pulpboard
Specifications
Capacity (Metric)
120 mL
Color
Amber
Diameter (Metric) Outer Neck
35 mm
Diameter (Metric) Inner Neck
25 mm
Height (Metric) With Closure
98 mm
Material
Glass (Type 3)
Quantity
24
Closure Color
White
Neck Finish
38-400
Closure Included
Yes
Mouth
Wide
Height (Metric)
95 mm
Product Type
Wide Mouth Packer Bottle
Closure Liner
Pulp/Vinyl
Closure Material
Polypropylene
Closure Size
38-400 mm
Diameter (Metric)
54 mm
Unit Size