Check Order Status
Surface Treated Sterile Tissue Culture Flasks, Plug Seal Cap
Actual product may vary
Fisherbrand
Surface Treated Sterile Tissue Culture Flasks, Plug Seal Cap
Perfect for cell growth.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Culture Area
FISFB012934
25 mL
12.5 cm
2
LSVFB012936
50 mL
25 cm
2
LSVFB012938
250 mL
75 cm
2
LSVFB012940
600 mL
182 cm
2
4 Options
Catalog number
FISFB012934
Price (MYR)
816.00
Add to cart
Capacity (Metric):
25 mL
Culture Area:
12.5 cm
2
Price (MYR)
816.00
Add to cart
Flat, free flask surface
Striation free to maximize usable growth area
Used in both open, closed systems
Angled neck design offers good pipet and cell scraper access
Upper triangular, wider base shape provides stability
Protruding ridge on back side of flask makes it easy for stacking
Special area near neck for easy mark writing
Both flask sides have engraved graduation
Sterilized by gamma irradiation
Non-pyrogenic
Specifications
Certifications/Compliance
CE
Closure Color
Blue
Color
Natural
Capacity (Metric)
25 mL
For Use With (Application)
Cell growth
Neck Style
Angled
No. per Case
200 Flasks
No. per Pack
10 Flasks
Product Type
Tissue Culture Flask
Surface Coating
Surface-treated
Writing Surface
Flat
Graduated
Yes
Sterility
Sterile
Bottom Shape
Flat
Closure Type
Plug Seal
Culture Area
12.5 cm
2
Unit Size