Autoclavable HD Biohazard Waste Bags
Autoclavable HD Biohazard Waste Bags
Actual product may vary
Fisher Catalogue
Autoclavable HD Biohazard Waste Bags
Strong and leak-resistant
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorDimensions (L x W)Thickness
FIS22-044562Red with Black Print19 x 14 in. (48 x 36 cm)1.25 mil
FIS22-04-4558Clear with Red Print24 x 12 in. (61 x 30 cm)1.50 mil
FIS22-044564Red with Black Print35 x 25 in. (89 x 64 cm)1.50 mil
FIS22-044565Red with Black Print36 x 45 in. (91 x 114 cm)1.50 mil
Catalog number FIS22-044562
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
Red with Black Print
Dimensions (L x W):
19 x 14 in. (48 x 36 cm)
Thickness:
1.25 mil
Strong and leak-resistant
  • Printed with biohazard symbol and text in English and Spanish
  • Supplied with twist tie closure
Specifications
Closure TypeTwist Tie
ColorRed with Black Print
Dimensions (L x W)19 x 14 in. (48 x 36 cm)
Length (Metric)48 cm
Width (Metric)36 cm
AutoclavableAutoclavable
IncludesBiohazard symbol and text in English and Spanish
Width (English)14 in.
Length (English)19 in.
Thickness1.25 mil
MaterialPolyethylene
Unit Size