Spilfyter™ KolorSafe™ Liquid Neutralizers for Bases
Spilfyter™ KolorSafe™ Liquid Neutralizers for Bases
Actual product may vary
Spilfyter™ KolorSafe™ Liquid Neutralizers for Bases
Spilfyter™ KolorSafe™ Liquid Neutralizers for Bases
Spilfyter™ KolorSafe™ Liquid Neutralizers for Bases
Take the guesswork out of neutralizing hazardous spills
Have Questions?
Catalog NumberCapacity (Metric)Packaging Quantity
FIS19-0218900.94 L12/Cs.
Catalog number FIS19-021890
Price (MYR)
2,740.00
CS
Add to cart
Capacity (Metric):
0.94 L
Packaging Quantity:
12/Cs.
Price (MYR)
2,740.00
CS
Add to cart
Unlike dry neutralizer, liquid reagent can be sprayed into hard-to-reach areas and on vertical surfaces.
  • Sodium-free liquid reagent changes color when neutralization is complete
  • Safe for use in laboratories and cleanrooms
Specifications
Capacity (English)0.24 gal.
Capacity (Metric)0.94 L
TypeBase Neutralizer
FormatLiquid
For Use With (Application)For lab and cleanrooms
IncludesColor change indicator when neutralized
Packaging Quantity12/Cs.
Shelf Life3 Years
Weight (English)31 lb.
Weight (Metric)14 kg
Unit SizeCS